missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H3342403

 View more versions of this product

Catalog No. AAH3342403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride
C8H4ClF3N4O2S
1g
Moisture sensitive
SADUDBJMXGGXDE-UHFFFAOYSA-N
1-[4-(trifluoromethyl)pyrimidin-2-yl]pyrazole-4-sulfonyl chloride
51063957
95%
1215564-15-0
MFCD11505058
UN3261
1-4-trifluoromethyl-2-pyrimidinyl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulphonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl pyrazole-4-sulfonyl chloride, chloro 1-4-trifluoromethyl pyrimidin-2-yl pyrazol-4-yl sulfone
C1=CN=C(N=C1C(F)(F)F)N2C=C(C=N2)S(=O)(=O)Cl
312.651
312.66

Chemical Identifiers

1215564-15-0
312.651
SADUDBJMXGGXDE-UHFFFAOYSA-N
51063957
C1=CN=C(N=C1C(F)(F)F)N2C=C(C=N2)S(=O)(=O)Cl
C8H4ClF3N4O2S
MFCD11505058
1-4-trifluoromethyl-2-pyrimidinyl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulphonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl pyrazole-4-sulfonyl chloride, chloro 1-4-trifluoromethyl pyrimidin-2-yl pyrazol-4-yl sulfone
1-[4-(trifluoromethyl)pyrimidin-2-yl]pyrazole-4-sulfonyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only