CAS RN 1215564-15-0
IUPAC Name:
1-[4-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride
Synonyms:
1-[4-(trifluoromethyl)pyrimidin-2-yl]pyrazole-4-sulfonyl chloride
Molecular Weight (g/mol):
312.65
Molecular Formula:
C8H4ClF3N4O2S
InChi Key:
SADUDBJMXGGXDE-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=NC(=NC=C1)N1C=C(C=N1)S(Cl)(=O)=O
1
–
1
of
1
results
1
Promotions Available
1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%, Thermo Scientific™
1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%, Thermo Scientific™
CAS: 1215564-15-0 Molecular Formula: C8H4ClF3N4O2S Molecular Weight (g/mol): 312.651 MDL Number: MFCD11505058 InChI Key: SADUDBJMXGGXDE-UHFFFAOYSA-N Synonym: 1-4-trifluoromethyl-2-pyrimidinyl-1h-pyrazole-4-sulfonyl chloride,1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulfonyl chloride,1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulphonyl chloride,1-4-trifluoromethyl pyrimidin-2-yl pyrazole-4-sulfonyl chloride,chloro 1-4-trifluoromethyl pyrimidin-2-yl pyrazol-4-yl sulfone PubChem CID: 51063957 IUPAC Name: 1-[4-(trifluoromethyl)pyrimidin-2-yl]pyrazole-4-sulfonyl chloride SMILES: C1=CN=C(N=C1C(F)(F)F)N2C=C(C=N2)S(=O)(=O)Cl