Learn More
1,4-Phenylene diisothiocyanate, 98%
CAS: 4044-65-9 | C8H4N2S2 | 192.254 g/mol
Supplier: Thermo Scientific Chemicals L0975606
Description
It is used in the treatment of hookworms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the treatment of hookworms.
Solubility
Hydrolyzes in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
| 1,4-Phenylene diisothiocyanate | |
| 128°C to 133°C | |
| C8H4N2S2 | |
| 5 g | |
| 2414340 | |
| 14,1308 | |
| Hydrolyzes in water. | |
| C1=CC(=CC=C1N=C=S)N=C=S | |
| 192.254 | |
| 192.26 |
| 4044-65-9 | |
| Irritating | |
| MFCD00004811 | |
| UN2923 | |
| Moisture sensitive | |
| bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate | |
| OMWQUXGVXQELIX-UHFFFAOYSA-N | |
| 1,4-diisothiocyanatobenzene | |
| 19958 | |
| 98% |
Chemical Identifiers
| 4044-65-9 | |
| 192.254 | |
| OMWQUXGVXQELIX-UHFFFAOYSA-N | |
| 19958 | |
| C1=CC(=CC=C1N=C=S)N=C=S |
| C8H4N2S2 | |
| MFCD00004811 | |
| bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate | |
| 1,4-diisothiocyanatobenzene |
Safety and Handling
GHS H Statement
H331-H334-H314-H318-H302-H312
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c
H301-H315-H319-H334-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 223-741-3
RTECSNumber : NX9150000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only