missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylene diisothiocyanate, 98%

Catalog No. p-7045193
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0975603 1 g
AAL0975606 5 g
AAL0975614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0975603 Supplier Thermo Scientific Chemicals Supplier No. L0975603

CAS: 4044-65-9 | C8H4N2S2 | 192.254 g/mol

It is used in the treatment of hookworms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the treatment of hookworms.

Solubility
Hydrolyzes in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4044-65-9
Molecular Formula C8H4N2S2
Molecular Weight (g/mol) 192.254
MDL Number MFCD00004811
InChI Key OMWQUXGVXQELIX-UHFFFAOYSA-N
Synonym bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate
PubChem CID 19958
IUPAC Name 1,4-diisothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)N=C=S

Specifications

Melting Point 128°C to 133°C
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2414340
Sensitivity Moisture sensitive
Merck Index 14,1308
Solubility Information Hydrolyzes in water.
Formula Weight 192.26
Percent Purity 98%
Chemical Name or Material 1,4-Phenylene diisothiocyanate
Show More Show Less
Hazard Category H301-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H314-H318-H302-H312
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 223-741-3
RTECSNumber NX9150000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only