missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Naphthoquinone, 99%, contains up to 6% water, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 128.5°C |
| Color | Khaki |
| Flash Point | 141°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Plastic bottle |
| Quantity | 500 g |
| Beilstein | 07, 724 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal if inhaled. May cause an allergic skin reaction. Toxic in contact with skin. Causes severe skin burns and eye damage. Toxic if swallowed. Very toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breat |
| EINECSNumber | 204-977-6 |
| RTECSNumber | QL7175000 |
| TSCA | TSCA |
RUO – Research Use Only