missing translation for 'onlineSavingsMsg'
Learn More

1,4-Naphthoquinone, 99%, contains up to 6% water

Catalog No. AC166711000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166711000 100 g Plastic bottle
AC166715000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC166711000 Supplier Thermo Scientific Chemicals Supplier No. 166711000

CAS: 130-15-4 | C10H6O2 | 158.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130-15-4 , 7732-18-5
Molecular Formula C10H6O2
Molecular Weight (g/mol) 158.16
MDL Number MFCD00001676
InChI Key FRASJONUBLZVQX-UHFFFAOYSA-N
Synonym 1,4-naphthoquinone, 1,4-naphthalenedione, p-naphthoquinone, naphthoquinone, alpha-naphthoquinone, 1,4-naphthylquinone, usaf cy-10, 1,4-dihydronaphthalene-1,4-dione, 1,4-dihydro-1,4-diketonaphthalene, 1,4-naftochinon
PubChem CID 8530
ChEBI CHEBI:27418
IUPAC Name naphthalene-1,4-dione
SMILES C1=CC=C2C(=O)C=CC(=O)C2=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.5°C
Color Khaki
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 07, 724
Fieser 01,714
Merck Index 15, 6480
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkali hydroxide solutions,freely soluble in hot alcohol,ether,benzene,,chloroform,carbon bisulfide and acetic acid
Formula Weight 158.16
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1, 4-Naphthoquinone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
May cause an allergic skin reaction.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breat
EINECSNumber 204-977-6
RTECSNumber QL7175000
TSCA TSCA

RUO – Research Use Only