missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Methylphenoxy)-2-propanol 90.0+%, TCI America™
$178.95 - $469.75
Chemical Identifiers
| CAS | 4369-08-8 |
|---|---|
| Molecular Formula | C10H14O2 |
| Molecular Weight (g/mol) | 166.22 |
| MDL Number | MFCD00191544 |
| InChI Key | VTOVIJQMFYQNSX-UHFFFAOYNA-N |
| Synonym | 1-(p-Tolyloxy)-2-propanol |
| PubChem CID | 12384823 |
| IUPAC Name | 1-(4-methylphenoxy)propan-2-ol |
| SMILES | CC1=CC=C(C=C1)OCC(C)O |
Chemical Identifiers
| 4369-08-8 | |
| 166.22 | |
| VTOVIJQMFYQNSX-UHFFFAOYNA-N | |
| 12384823 | |
| CC1=CC=C(C=C1)OCC(C)O |
| C10H14O2 | |
| MFCD00191544 | |
| 1-(p-Tolyloxy)-2-propanol | |
| 1-(4-methylphenoxy)propan-2-ol |
Specifications
| 4369-08-8 | |
| 130°C | |
| MFCD00191544 | |
| 1-(p-Tolyloxy)-2-propanol | |
| CC1=CC=C(C=C1)OCC(C)O | |
| 166.22 | |
| 166.22 | |
| Crystalline Lumps |
| White-Yellow | |
| C10H14O2 | |
| 5 g | |
| VTOVIJQMFYQNSX-UHFFFAOYNA-N | |
| 1-(4-methylphenoxy)propan-2-ol | |
| 12384823 | |
| ≥90.0% (GC) | |
| 1-(4-Methylphenoxy)-2-propanol |
Safety and Handling
TSCA : No