missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)-1-cyclopropanecarbonitrile, 96%

Catalog No. AC170620250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170620250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC170620250 Supplier Thermo Scientific Chemicals Supplier No. 170620250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16728-00-0 | C11H11NO | 173.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16728-00-0
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.22
MDL Number MFCD00019212
InChI Key OIQRGAQXQLASRC-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl cyclopropanecarbonitrile, 1-4-methoxyphenyl cyclopropane-1-carbonitrile, cyclopropanecarbonitrile, 1-4-methoxyphenyl, 1-4-methoxyphenyl-1-cyclopropanecarbonitrile, 1-4-methoxy-phenyl cyclopropanecarbonitrile, 4-1-cyanocyclopropyl anisole, 1-4-methoxy-phenyl-cyclopropanecarbonitrile, cyclopropanecarbonitrile,1-4-methoxyphenyl, 1-4-methoxyphenyl-cyclopropane-1-carbonitrile, 1-4-methoxyphenyl cyclopropane carbonitrile
PubChem CID 85574
IUPAC Name 1-(4-methoxyphenyl)cyclopropane-1-carbonitrile
SMILES COC1=CC=C(C=C1)C1(CC1)C#N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0800g/mL
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5415 to 1.5435
Quantity 25 g
Specific Gravity 1.08
Formula Weight 173.21
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclopropanecarbonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 240-786-4

RUO – Research Use Only