accessibility menu, dialog, popup

UserName

CAS RN 16728-00-0

IUPAC Name: 1-(4-methoxyphenyl)cyclopropane-1-carbonitrile
Synonyms: 1-(4-methoxyphenyl)cyclopropane-1-carbonitrile
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: OIQRGAQXQLASRC-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1(CC1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results