missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Hydroxyphenyl)piperazine 98.0+%, TCI America™

Catalog No. H089125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H089125G 25 g
H0891250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H089125G Supplier TCI America Supplier No. H089125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56621-48-8
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.24
MDL Number MFCD00066156
InChI Key GPEOAEVZTOQXLG-UHFFFAOYSA-N
Synonym 1-4-hydroxyphenyl piperazine, 4-piperazin-1-yl phenol, 4-1-piperazinyl phenol, n-4-hydroxyphenyl piperazine, p-1-piperazinyl phenol, 4-piperazinophenol, phenol, 4-1-piperazinyl, 1-4-hydroxyphenyl-piperazine, 4-piperazinylphenol, 4-piperazinyl phenol
PubChem CID 92467
IUPAC Name 4-(piperazin-1-yl)phenol
SMILES OC1=CC=C(C=C1)N1CCNCC1

Specifications

Melting Point 223°C
Quantity 25 g
Formula Weight 178.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Hydroxyphenyl)piperazine