missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole 90.0+%, TCI America™
Supplier: TCI America H089625G
Specifications
| 1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole | |
| 162°C | |
| C7H6N4OS | |
| 25 g | |
| 1-4-hydroxyphenyl-5-mercaptotetrazole, 1-4-hydroxyphenyl-1h-tetrazole-5-thiol, 1-4-hydroxyphenyl-2h-tetrazole-5-thione, 1-4-hydroxyphenyl-5-mercapto-1h-tetrazole, 5h-tetrazole-5-thione, 1,2-dihydro-1-4-hydroxyphenyl, 1-4-hydroxyphenyl-1h-tetrazole-5 4h-thione, 4-5-sulfanyl-1h-1,2,3,4-tetrazol-1-yl phenol, 1,2-dihydro-1-p-hydroxyphenyl-5h-tetrazole-5-thione, 4-5-mercapto-1h-tetrazol-1-yl phenol, 4-5-sulfanyl-1h-tetrazol-1-yl phenol | |
| C1=CC(=CC=C1N2C(=S)N=NN2)O | |
| 194.212 | |
| 194.21 | |
| Crystalline Powder |
| 52431-78-4 | |
| White-Yellow | |
| MFCD00132898 | |
| 1325 | |
| MOXZSKYLLSPATM-UHFFFAOYSA-N | |
| 1-(4-hydroxyphenyl)-2H-tetrazole-5-thione | |
| 3034725 | |
| ≥90.0% (T) |
Chemical Identifiers
| 52431-78-4 | |
| 194.212 | |
| MOXZSKYLLSPATM-UHFFFAOYSA-N | |
| 3034725 | |
| C1=CC(=CC=C1N2C(=S)N=NN2)O |
| C7H6N4OS | |
| MFCD00132898 | |
| 1-4-hydroxyphenyl-5-mercaptotetrazole, 1-4-hydroxyphenyl-1h-tetrazole-5-thiol, 1-4-hydroxyphenyl-2h-tetrazole-5-thione, 1-4-hydroxyphenyl-5-mercapto-1h-tetrazole, 5h-tetrazole-5-thione, 1,2-dihydro-1-4-hydroxyphenyl, 1-4-hydroxyphenyl-1h-tetrazole-5 4h-thione, 4-5-sulfanyl-1h-1,2,3,4-tetrazol-1-yl phenol, 1,2-dihydro-1-p-hydroxyphenyl-5h-tetrazole-5-thione, 4-5-mercapto-1h-tetrazol-1-yl phenol, 4-5-sulfanyl-1h-tetrazol-1-yl phenol | |
| 1-(4-hydroxyphenyl)-2H-tetrazole-5-thione |
Safety and Handling
EINECSNumber : (5)-6069
TSCA : Yes