missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole 90.0+%, TCI America™

Catalog No. p-7130791
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08965G 5 g
H089625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08965G Supplier TCI America Supplier No. H08965G
Only null left

Chemical Identifiers

CAS 52431-78-4
Molecular Formula C7H6N4OS
Molecular Weight (g/mol) 194.212
MDL Number MFCD00132898
InChI Key MOXZSKYLLSPATM-UHFFFAOYSA-N
Synonym 1-4-hydroxyphenyl-5-mercaptotetrazole, 1-4-hydroxyphenyl-1h-tetrazole-5-thiol, 1-4-hydroxyphenyl-2h-tetrazole-5-thione, 1-4-hydroxyphenyl-5-mercapto-1h-tetrazole, 5h-tetrazole-5-thione, 1,2-dihydro-1-4-hydroxyphenyl, 1-4-hydroxyphenyl-1h-tetrazole-5 4h-thione, 4-5-sulfanyl-1h-1,2,3,4-tetrazol-1-yl phenol, 1,2-dihydro-1-p-hydroxyphenyl-5h-tetrazole-5-thione, 4-5-mercapto-1h-tetrazol-1-yl phenol, 4-5-sulfanyl-1h-tetrazol-1-yl phenol
PubChem CID 3034725
IUPAC Name 1-(4-hydroxyphenyl)-2H-tetrazole-5-thione
SMILES C1=CC(=CC=C1N2C(=S)N=NN2)O

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 5 g
UN Number 1325
Formula Weight 194.21
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole
EINECSNumber (5)-6069
TSCA Yes