missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dinitrobenzene, 98%

Catalog No. AC408650050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408650050 5 g Glass bottle
AC408650250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC408650050 Supplier Thermo Scientific Chemicals Supplier No. 408650050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100-25-4 | C6H4N2O4 | 168.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-25-4
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.11
MDL Number MFCD00007314
InChI Key FYFDQJRXFWGIBS-UHFFFAOYSA-N
Synonym p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene
PubChem CID 7492
ChEBI CHEBI:51398
IUPAC Name 1,4-dinitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 173°C to 175°C
Color Ochre to Orange
Density 1.6300g/mL
Boiling Point 299°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula NO2C6H4NO2
Quantity 5 g
Beilstein 05, 261
Specific Gravity 1.63
Merck Index 15, 3299
Solubility Information Solubility in water: 0.8g/L (20°C). Other solubilities: soluble in ethanol,ether,benzene,chloroform,,ethyl acetate,acetone and toluene
Formula Weight 168.11
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 1, 4-Dinitrobenzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Fatal in contact with skin.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or do
EINECSNumber 202-833-7
RTECSNumber CZ7525000
TSCA TSCA

RUO – Research Use Only