Learn More
1,4-Dinitrobenzene, 98+%
CAS: 100-25-4 | C6H4N2O4 | 168.108 g/mol
Supplier: Thermo Scientific Chemicals A1029314
Description
1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.
Solubility
Soluble in water. (0.8 g/L) at 20°C.
Notes
Incompatible with bases, alkali metals, oxidizing agents and nitric acid.
Specifications
| 1,4-Dinitrobenzene | |
| 172°C to 176°C | |
| 299°C | |
| C6H4N2O4 | |
| 25 g | |
| 1105828 | |
| p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene | |
| FYFDQJRXFWGIBS-UHFFFAOYSA-N | |
| 1,4-dinitrobenzene | |
| 7492 | |
| 168.11 |
| 100-25-4 | |
| 1.625 | |
| 150°C (302°F) | |
| MFCD00007314 | |
| UN3443 | |
| 14,3273 | |
| Soluble in water. (0.8g/L) at 20°C. | |
| C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-] | |
| 168.108 | |
| CHEBI:51398 | |
| ≥98% |
Chemical Identifiers
| 100-25-4 | |
| 168.108 | |
| FYFDQJRXFWGIBS-UHFFFAOYSA-N | |
| 7492 | |
| 1,4-dinitrobenzene |
| C6H4N2O4 | |
| MFCD00007314 | |
| p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene | |
| CHEBI:51398 | |
| C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-] |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DINITROBENZENES, SOLID
EINECSNumber : 202-833-7
RTECSNumber : CZ7525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only