missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dinitrobenzene, 98+%

Catalog No. AAA1029306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1029306 5 g
AAA1029314 25 g
AAA1029322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1029306 Supplier Thermo Scientific Chemicals Supplier No. A1029306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100-25-4 | C6H4N2O4 | 168.108 g/mol

1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

Solubility
Soluble in water. (0.8 g/L) at 20°C.

Notes
Incompatible with bases, alkali metals, oxidizing agents and nitric acid.

Chemical Identifiers

CAS 100-25-4
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
MDL Number MFCD00007314
InChI Key FYFDQJRXFWGIBS-UHFFFAOYSA-N
Synonym p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene
PubChem CID 7492
ChEBI CHEBI:51398
IUPAC Name 1,4-dinitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 172°C to 176°C
Density 1.625
Boiling Point 299°C
Flash Point 150°C (302°F)
Quantity 5 g
UN Number UN3443
Beilstein 1105828
Merck Index 14,3273
Solubility Information Soluble in water. (0.8g/L) at 20°C.
Formula Weight 168.11
Percent Purity ≥98%
Chemical Name or Material 1,4-Dinitrobenzene
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DINITROBENZENES, SOLID
EINECSNumber 202-833-7
RTECSNumber CZ7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only