Learn More
1,4-Diiodobutane, 99%, stab. with copper
CAS: 628-21-7 | C4H8I2 | 309.92 g/mol
Supplier: Thermo Scientific Chemicals A1621622
Description
1,4-Diiodobutane is used as an additive to enhance perovskite solar cell performance. It is used as a crosslinking agent for the synthesis of anion exchange membranes from cellulose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Diiodobutane is used as an additive to enhance perovskite solar cell performance. It is used as a crosslinking agent for the synthesis of anion exchange membranes from cellulose.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agent.
Specifications
| 1,4-Diiodobutane | |
| 628-21-7 | |
| 2.35 | |
| C4H8I2 | |
| MFCD00001099 | |
| 1098276 | |
| tetramethylene iodide, butane, 1,4-diiodo, tetramethylene diiodide, 1,4-diodobutane, a,?-diiodobutane, 1,4 diiodobutane, butane,4-diiodo, 1,4-diiodo-butane, pubchem3842, 1,4-bis iodanyl butane | |
| ROUYUBHVBIKMQO-UHFFFAOYSA-N | |
| 1,4-diiodobutane | |
| 12337 | |
| 99% |
| stabilized with copper | |
| 5°C to 7°C | |
| 108°C to 110°C (10 mmHg) | |
| 1.6212 | |
| 100 g | |
| Light sensitive | |
| Insoluble in water. | |
| ICCCCI | |
| 309.92 | |
| 309.92 |
Chemical Identifiers
| 628-21-7 | |
| 309.92 | |
| ROUYUBHVBIKMQO-UHFFFAOYSA-N | |
| 12337 | |
| ICCCCI |
| C4H8I2 | |
| MFCD00001099 | |
| tetramethylene iodide, butane, 1,4-diiodo, tetramethylene diiodide, 1,4-diodobutane, a,?-diiodobutane, 1,4 diiodobutane, butane,4-diiodo, 1,4-diiodo-butane, pubchem3842, 1,4-bis iodanyl butane | |
| 1,4-diiodobutane |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 211-032-1
RTECSNumber : EJ9200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only