missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diiodobutane, 99%, stab. with copper

Catalog No. p-7023130
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1621614 25 g
AAA1621622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1621614 Supplier Thermo Scientific Chemicals Supplier No. A1621614
Only null left

CAS: 628-21-7 | C4H8I2 | 309.92 g/mol

1,4-Diiodobutane is used as an additive to enhance perovskite solar cell performance. It is used as a crosslinking agent for the synthesis of anion exchange membranes from cellulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diiodobutane is used as an additive to enhance perovskite solar cell performance. It is used as a crosslinking agent for the synthesis of anion exchange membranes from cellulose.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agent.

Chemical Identifiers

CAS 628-21-7
Molecular Formula C4H8I2
Molecular Weight (g/mol) 309.92
MDL Number MFCD00001099
InChI Key ROUYUBHVBIKMQO-UHFFFAOYSA-N
Synonym tetramethylene iodide, butane, 1,4-diiodo, tetramethylene diiodide, 1,4-diodobutane, a,?-diiodobutane, 1,4 diiodobutane, butane,4-diiodo, 1,4-diiodo-butane, pubchem3842, 1,4-bis iodanyl butane
PubChem CID 12337
IUPAC Name 1,4-diiodobutane
SMILES ICCCCI

Specifications

Melting Point 5°C to 7°C
Density 2.35
Boiling Point 108°C to 110°C (10 mmHg)
Refractive Index 1.6212
Quantity 25 g
Beilstein 1098276
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 309.92
Percent Purity 99%
Chemical Name or Material 1,4-Diiodobutane, stabilized with copper
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 211-032-1
RTECSNumber EJ9200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only