missing translation for 'onlineSavingsMsg'
Learn More

1,4-Difluorobenzene 99.0+%, TCI America™

Catalog No. p-7126834
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16285G 5 g
D162825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16285G Supplier TCI America Supplier No. D16285G
Only null left

Chemical Identifiers

CAS 540-36-3
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000344
InChI Key QUGUFLJIAFISSW-UHFFFAOYSA-N
Synonym p-difluorobenzene, benzene, 1,4-difluoro, para-difluorobenzene, benzene, p-difluoro, 2,5-difluorobenzene, unii-y9fq2ef0z6, 1,4-difluorbenzol, y9fq2ef0z6, benzene,4-difluoro, pubchem2309
PubChem CID 10892
ChEBI CHEBI:38585
IUPAC Name 1,4-difluorobenzene
SMILES FC1=CC=C(F)C=C1

Specifications

Melting Point -13°C
Color Yellow
Boiling Point 88°C
Quantity 5 g
UN Number 1993
Formula Weight 114.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Difluorobenzene
EINECSNumber (3)-2688
RTECSNumber CZ5658000
TSCA No