accessibility menu, dialog, popup

UserName

FF

CAS RN 540-36-3

IUPAC Name: 1,4-difluorobenzene
Synonyms: 1,4-difluorobenzene P-difluorobenzene
Molecular Weight (g/mol): 114.1
Molecular Formula: C6H4F2
InChi Key: QUGUFLJIAFISSW-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)