missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethoxybenzene, 98%

Catalog No. p-7022551
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1538514 25 g
AAA1538522 100 g
AAA1538536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1538514 Supplier Thermo Scientific Chemicals Supplier No. A1538514

missing translation for 'validMainframeMsg'

CAS: 122-95-2 | C10H14O2 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-95-2
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00015146
InChI Key VWGNFIQXBYRDCH-UHFFFAOYSA-N
Synonym benzene, 1,4-diethoxy, p-diethoxybenzene, hydroquinone diethyl ether, benzene, p-diethoxy, diethoxybenzene, 1,4-diethoxy-benzen, benzene,4-diethoxy, 1,4-diethoxybenzol, hydroquinonedlethylether, acmc-209ans
PubChem CID 67150
IUPAC Name 1,4-diethoxybenzene
SMILES CCOC1=CC=C(OCC)C=C1

Specifications

Melting Point 69°C to 71°C
Boiling Point 245°C to 248°C
Quantity 25 g
Beilstein 2044373
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 1,4-Diethoxybenzene
EINECSNumber 204-585-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only