missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dicyclohexylbenzene 98.0+%, TCI America™
$197.59 - $1038.29
Chemical Identifiers
| CAS | 1087-02-1 |
|---|---|
| Molecular Formula | C18H26 |
| Molecular Weight (g/mol) | 242.406 |
| MDL Number | MFCD00001452 |
| InChI Key | QQFSIGWYINAJOB-UHFFFAOYSA-N |
| Synonym | p-dicyclohexylbenzene, unii-a4m5u01mcy, a4m5u01mcy, benzene, 1,4-dicyclohexyl, p-dicyclohexylbenzol, benzene,4-dicyclohexyl, benzene, p-dicyclohexyl, p-dicyclohexyl benzene, qqfsigwyinajob-uhfffaoysa, unii-n5v9zbl13v component |
| PubChem CID | 70664 |
| IUPAC Name | 1,4-dicyclohexylbenzene |
| SMILES | C1CCC(CC1)C2=CC=C(C=C2)C3CCCCC3 |
Chemical Identifiers
| 1087-02-1 | |
| 242.406 | |
| QQFSIGWYINAJOB-UHFFFAOYSA-N | |
| 70664 | |
| C1CCC(CC1)C2=CC=C(C=C2)C3CCCCC3 |
| C18H26 | |
| MFCD00001452 | |
| p-dicyclohexylbenzene, unii-a4m5u01mcy, a4m5u01mcy, benzene, 1,4-dicyclohexyl, p-dicyclohexylbenzol, benzene,4-dicyclohexyl, benzene, p-dicyclohexyl, p-dicyclohexyl benzene, qqfsigwyinajob-uhfffaoysa, unii-n5v9zbl13v component | |
| 1,4-dicyclohexylbenzene |
Specifications
| 1087-02-1 | |
| White | |
| C18H26 | |
| 25 g | |
| QQFSIGWYINAJOB-UHFFFAOYSA-N | |
| 1,4-dicyclohexylbenzene | |
| 70664 | |
| ≥98.0% (GC) | |
| 1,4-Dicyclohexylbenzene |
| 102°C | |
| 195°C | |
| MFCD00001452 | |
| p-dicyclohexylbenzene, unii-a4m5u01mcy, a4m5u01mcy, benzene, 1,4-dicyclohexyl, p-dicyclohexylbenzol, benzene,4-dicyclohexyl, benzene, p-dicyclohexyl, p-dicyclohexyl benzene, qqfsigwyinajob-uhfffaoysa, unii-n5v9zbl13v component | |
| C1CCC(CC1)C2=CC=C(C=C2)C3CCCCC3 | |
| 242.406 | |
| 242.41 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0041
TSCA : No