CAS RN 1087-02-1
IUPAC Name:
1,4-dicyclohexylbenzene
Synonyms:
1,4-dicyclohexylbenzene
Molecular Weight (g/mol):
242.41
Molecular Formula:
C18H26
InChi Key:
QQFSIGWYINAJOB-UHFFFAOYSA-N
SMILES:
C1CCC(CC1)C1=CC=C(C=C1)C1CCCCC1
1
–
1
of
1
results
1
1,4-Dicyclohexylbenzene 98.0+%, TCI America™
CAS: 1087-02-1 Molecular Formula: C18H26 Molecular Weight (g/mol): 242.406 MDL Number: MFCD00001452 InChI Key: QQFSIGWYINAJOB-UHFFFAOYSA-N Synonym: p-dicyclohexylbenzene,unii-a4m5u01mcy,a4m5u01mcy,benzene, 1,4-dicyclohexyl,p-dicyclohexylbenzol,benzene,4-dicyclohexyl,benzene, p-dicyclohexyl,p-dicyclohexyl benzene,qqfsigwyinajob-uhfffaoysa,unii-n5v9zbl13v component PubChem CID: 70664 IUPAC Name: 1,4-dicyclohexylbenzene SMILES: C1CCC(CC1)C2=CC=C(C=C2)C3CCCCC3