missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dicyclohexylbenzene 98.0+%, TCI America™
Supplier: TCI America D1028250G
Specifications
| 1,4-Dicyclohexylbenzene | |
| 102°C | |
| 195°C | |
| MFCD00001452 | |
| p-dicyclohexylbenzene, unii-a4m5u01mcy, a4m5u01mcy, benzene, 1,4-dicyclohexyl, p-dicyclohexylbenzol, benzene,4-dicyclohexyl, benzene, p-dicyclohexyl, p-dicyclohexyl benzene, qqfsigwyinajob-uhfffaoysa, unii-n5v9zbl13v component | |
| C1CCC(CC1)C2=CC=C(C=C2)C3CCCCC3 | |
| 242.406 | |
| 242.41 | |
| Crystalline Powder |
| 1087-02-1 | |
| White | |
| C18H26 | |
| 250 g | |
| QQFSIGWYINAJOB-UHFFFAOYSA-N | |
| 1,4-dicyclohexylbenzene | |
| 70664 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1087-02-1 | |
| 242.406 | |
| QQFSIGWYINAJOB-UHFFFAOYSA-N | |
| 70664 | |
| C1CCC(CC1)C2=CC=C(C=C2)C3CCCCC3 |
| C18H26 | |
| MFCD00001452 | |
| p-dicyclohexylbenzene, unii-a4m5u01mcy, a4m5u01mcy, benzene, 1,4-dicyclohexyl, p-dicyclohexylbenzol, benzene,4-dicyclohexyl, benzene, p-dicyclohexyl, p-dicyclohexyl benzene, qqfsigwyinajob-uhfffaoysa, unii-n5v9zbl13v component | |
| 1,4-dicyclohexylbenzene |
Safety and Handling
EINECSNumber : (4)-0041
TSCA : No