missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorobutane 98.0+%, TCI America™

Supplier:  TCI America D0349500ML

 View more versions of this product

Catalog No. D0349500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,4-Dichlorobutane
Colorless
C4H8Cl2
500 mL
butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc
ClCCCCCl
127.01
127.01
Liquid
110-56-5
155°C
MFCD00001011
1993
KJDRSWPQXHESDQ-UHFFFAOYSA-N
1,4-dichlorobutane
8059
≥98.0% (GC)

Chemical Identifiers

110-56-5
127.01
KJDRSWPQXHESDQ-UHFFFAOYSA-N
8059
ClCCCCCl
C4H8Cl2
MFCD00001011
butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc
1,4-dichlorobutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0061

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions