missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorobutane 98.0+%, TCI America™

Catalog No. p-7126164
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
D034925ML 25 mL
D0349500ML 500 mL
2 options

Catalog No. D034925ML

Supplier: TCI America D034925ML

Only null left

Chemical Identifiers

CAS 110-56-5
Molecular Formula C4H8Cl2
Molecular Weight (g/mol) 127.01
MDL Number MFCD00001011
InChI Key KJDRSWPQXHESDQ-UHFFFAOYSA-N
Synonym butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc
PubChem CID 8059
IUPAC Name 1,4-dichlorobutane
SMILES ClCCCCCl

Specifications

Color Colorless
Boiling Point 155°C
Quantity 25 mL
UN Number 1993
Formula Weight 127.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dichlorobutane
EINECSNumber (2)-0061
TSCA Yes