Learn More
1,4-Dichlorobenzene, 99+%
CAS: 106-46-7 | C6H4Cl2 | 146.998 g/mol
Supplier: Thermo Scientific Chemicals A1629036
Description
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, aluminum and its alloys, some plastics.
Specifications
| 1,4-Dichlorobenzene | |
| 52°C to 56°C | |
| 173°C to 174°C | |
| Strong | |
| MFCD00000604 | |
| UN3077 | |
| 14,3057 | |
| It is insoluble in water. | |
| C1=CC(=CC=C1Cl)Cl | |
| 146.998 | |
| CHEBI:28618 | |
| ≥99% |
| 106-46-7 | |
| 1.248 | |
| 65°C (149°F) | |
| C6H4Cl2 | |
| 500 g | |
| 1680023 | |
| p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola | |
| OCJBOOLMMGQPQU-UHFFFAOYSA-N | |
| 1,4-dichlorobenzene | |
| 4685 | |
| 147 |
Chemical Identifiers
| 106-46-7 | |
| 146.998 | |
| OCJBOOLMMGQPQU-UHFFFAOYSA-N | |
| 4685 | |
| 1,4-dichlorobenzene |
| C6H4Cl2 | |
| MFCD00000604 | |
| p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola | |
| CHEBI:28618 | |
| C1=CC(=CC=C1Cl)Cl |
Safety and Handling
GHS H Statement
H351-H302-H332-H319
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
H319-H350
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 203-400-5
RTECSNumber : CZ4550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only