missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorobenzene, 99+%

Catalog No. p-7023164
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1629022 100 g
AAA1629036 500 g
AAA162900E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1629022 Supplier Thermo Scientific Chemicals Supplier No. A1629022
Only null left

CAS: 106-46-7 | C6H4Cl2 | 146.998 g/mol

1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, aluminum and its alloys, some plastics.

Chemical Identifiers

CAS 106-46-7
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000604
InChI Key OCJBOOLMMGQPQU-UHFFFAOYSA-N
Synonym p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
PubChem CID 4685
ChEBI CHEBI:28618
IUPAC Name 1,4-dichlorobenzene
SMILES C1=CC(=CC=C1Cl)Cl

Specifications

Melting Point 52°C to 56°C
Density 1.248
Boiling Point 173°C to 174°C
Flash Point 65°C (149°F)
Odor Strong
Quantity 100 g
UN Number UN3077
Beilstein 1680023
Merck Index 14,3057
Solubility Information It is insoluble in water.
Formula Weight 147
Percent Purity ≥99%
Chemical Name or Material 1,4-Dichlorobenzene
Show More Show Less
Hazard Category H319-H350
Hazard Statement GHS H Statement
H351-H302-H332-H319
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 203-400-5
RTECSNumber CZ4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only