accessibility menu, dialog, popup

UserName

CAS RN 106-46-7

IUPAC Name: 1,4-dichlorobenzene
Synonyms: paradi paradichlorobenzene preparation caswell no. 632 parazene p-dcb para-DCB paradow p-diclorobenzene PDB paradichlorbenzol
Molecular Weight (g/mol): 147
Molecular Formula: C6H4Cl2
InChi Key: OCJBOOLMMGQPQU-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)