missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromotetrafluorobenzene, 99%

Catalog No. p-7024008
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1813503 1 g
AAA1813506 5 g
AAA1813514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1813503 Supplier Thermo Scientific Chemicals Supplier No. A1813503
Only null left

CAS: 344-03-6 | C6Br2F4 | 307.87 g/mol

It is used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a chemical intermediate.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 344-03-6
Molecular Formula C6Br2F4
Molecular Weight (g/mol) 307.87
MDL Number MFCD00000309
InChI Key QFTZULJNRAHOIY-UHFFFAOYSA-N
Synonym 1,4-dibromotetrafluorobenzene, p-dibromotetrafluorobenzene, benzene, 1,4-dibromo-2,3,5,6-tetrafluoro, benzene, 1,4-dibromotetrafluoro, 1,3,5,6-tetrafluorobenzene, benzene,4-dibromotetrafluoro, emolecules 536967, tetrafluoro-1,4-dibromo benzene, perfluoro-1,4-dibromobenzene
PubChem CID 67653
IUPAC Name 1,4-dibromo-2,3,5,6-tetrafluorobenzene
SMILES FC1=C(F)C(Br)=C(F)C(F)=C1Br

Specifications

Melting Point 78°C to 81°C
Quantity 1 g
Formula Weight 307.87
Percent Purity 99%
Chemical Name or Material 1,4-Dibromotetrafluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-448-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only