missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromopentane 95.0+%, TCI America™

Catalog No. p-7126718
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14085G 5 g
D140825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14085G Supplier TCI America Supplier No. D14085G
Only null left

Chemical Identifiers

CAS 626-87-9
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.943
MDL Number MFCD00000153
InChI Key CNBFRBXEGGRSPL-UHFFFAOYSA-N
Synonym pentane, 1,4-dibromo, 1,4-dibrompentan, pentane,4-dibromo, 1,4-dibromo-pentane, pubchem12546, 1,4-dibromopentane, hexanoic acid monoethyl ester
PubChem CID 79082
IUPAC Name 1,4-dibromopentane
SMILES CC(CCCBr)Br

Specifications

Color Yellow
Boiling Point 202°C
Quantity 5 g
Formula Weight 229.94
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dibromopentane
TSCA Yes
Recommended Storage Refrigerator