accessibility menu, dialog, popup

UserName

H3CBrBr

CAS RN 626-87-9

IUPAC Name: 1,4-dibromopentane
Synonyms: pentane, 1,4-dibromo-
Molecular Weight (g/mol): 229.94
Molecular Formula: C5H10Br2
InChi Key: CNBFRBXEGGRSPL-UHFFFAOYNA-N
SMILES: CC(Br)CCCBr
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)