missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromopentane, 97%

Catalog No. AAL0984214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0984214 25 g
AAL0984222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0984214 Supplier Thermo Scientific Chemicals Supplier No. L0984214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-87-9 | C5H10Br2 | 229.943 g/mol

1,4-Dibromopentane is used in the preparation of pharaoh ant trail pheromone stereoisomer, indolizidine alkaloid. It is also involved in the preparation 1,2-dimethyl-1-phenylcyclopentane and boc-L-2-amino-6-bromoheptanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromopentane is used in the preparation of pharaoh ant trail pheromone stereoisomer, indolizidine alkaloid. It is also involved in the preparation 1,2-dimethyl-1-phenylcyclopentane and boc-L-2-amino-6-bromoheptanoic acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 626-87-9
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.943
MDL Number MFCD00000153
InChI Key CNBFRBXEGGRSPL-UHFFFAOYSA-N
Synonym pentane, 1,4-dibromo, 1,4-dibrompentan, pentane,4-dibromo, 1,4-dibromo-pentane, pubchem12546, 1,4-dibromopentane, hexanoic acid monoethyl ester
PubChem CID 79082
IUPAC Name 1,4-dibromopentane
SMILES CC(CCCBr)Br

Specifications

Melting Point -34°C
Density 1.687
Boiling Point 102°C to 103°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5085
Quantity 25 g
Beilstein 605295
Solubility Information Insoluble in water.
Formula Weight 229.95
Percent Purity 97%
Chemical Name or Material 1,4-Dibromopentane
Show More Show Less
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-967-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only