missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromobenzene, 98%

CAS: 106-37-6 | C6H4Br2 | 235.91 g/mol

Supplier:  Thermo Scientific Chemicals A1051722

 View more versions of this product

Catalog No. AAA1051722


Only null left
Explore available promotions

Description

Description

1,4-Dibromobenzene is used in the synthesis of 2,5-dibromoacetophenone, triarylamines and trans- stilbenes. It is widely used as a heavy liquid solvent as well as a motor oil additive. It finds application as an intermediate in the manufacture of organic chemicals such as dyes, pharmaceuticals and flame retardants for polymeric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromobenzene is used in the synthesis of 2,5-dibromoacetophenone, triarylamines and trans- stilbenes. It is widely used as a heavy liquid solvent as well as a motor oil additive. It finds application as an intermediate in the manufacture of organic chemicals such as dyes, pharmaceuticals and flame retardants for polymeric materials.

Solubility
Soluble in alcohol, benzene, chloroform, toluene and carbon tetrachloride. Insoluble in ether.

Notes
Avoid excessive heat and light. Incompatible with strong oxidizing agents.
Specifications

Specifications

1, 4-Dibromobenzene
86°C to 90°C
219°C to 220°C
98%
1.5742
100 g
1904543
p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071
SWJPEBQEEAHIGZ-UHFFFAOYSA-N
1,4-dibromobenzene
7804
235.91
106-37-6
1.841 g/mL
>100°C (212°F)
C6H4Br2
MFCD00000089
UN3077
14,3019
Soluble in alcohol,benzene,chloroform,toluene and carbon tetrachloride. Insoluble in ether.
BrC1=CC=C(Br)C=C1
235.91
CHEBI:37150
98%

Chemical Identifiers

106-37-6
235.91
SWJPEBQEEAHIGZ-UHFFFAOYSA-N
7804
1,4-dibromobenzene
C6H4Br2
MFCD00000089
p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071
CHEBI:37150
BrC1=CC=C(Br)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 203-390-2

RTECSNumber : CZ1791000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only