missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromobenzene, 98%

Catalog No. AAA1051718
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
100 g
250 g
2500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1051718 50 g
AAA1051722 100 g
AAA1051730 250 g
AAA105170E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Catalog No. AAA1051718 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' A1051718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 106-37-6 | C6H4Br2 | 235.91 g/mol

1,4-Dibromobenzene is used in the synthesis of 2,5-dibromoacetophenone, triarylamines and trans- stilbenes. It is widely used as a heavy liquid solvent as well as a motor oil additive. It finds application as an intermediate in the manufacture of organic chemicals such as dyes, pharmaceuticals and flame retardants for polymeric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromobenzene is used in the synthesis of 2,5-dibromoacetophenone, triarylamines and trans- stilbenes. It is widely used as a heavy liquid solvent as well as a motor oil additive. It finds application as an intermediate in the manufacture of organic chemicals such as dyes, pharmaceuticals and flame retardants for polymeric materials.

Solubility
Soluble in alcohol, benzene, chloroform, toluene and carbon tetrachloride. Insoluble in ether.

Notes
Avoid excessive heat and light. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 106-37-6
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000089
InChI Key SWJPEBQEEAHIGZ-UHFFFAOYSA-N
Synonym p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071
PubChem CID 7804
ChEBI CHEBI:37150
IUPAC Name 1,4-dibromobenzene
SMILES BrC1=CC=C(Br)C=C1

Specifications

Melting Point 86°C to 90°C
Density 1.841
Boiling Point 219°C to 220°C
Flash Point >100°C (212°F)
Odor Characteristic
Assay Percent Range 98%
Refractive Index 1.5742
Quantity 50 g
UN Number UN3077
Beilstein 1904543
Merck Index 14,3019
Solubility Information Soluble in alcohol,benzene,chloroform,toluene and carbon tetrachloride. Insoluble in ether.
Formula Weight 235.91
Percent Purity 98%
Chemical Name or Material 1,4-Dibromobenzene
Show More Show Less
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 203-390-2
RTECSNumber CZ1791000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only