missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromo-2,5-dimethoxybenzene 98.0+%, TCI America™
Supplier: TCI America D39665G
Specifications
| 1,4-Dibromo-2,5-dimethoxybenzene | |
| 147°C | |
| C8H8Br2O2 | |
| 5 g | |
| CHCLRVOURKGRSW-UHFFFAOYSA-N | |
| 1,4-dibromo-2,5-dimethoxybenzene | |
| 231240 | |
| ≥98.0% (GC) |
| 2674-34-2 | |
| White-Yellow | |
| MFCD00025694 | |
| 2,5-dibromo-1,4-dimethoxybenzene, 1,4-dibromo-2,5-dimethoxy-benzene, 2,5-dibromohydroquinone dimethyl ether, benzene, 1,4-dibromo-2,5-dimethoxy, pubchem16840, acmc-209gsv, maybridge1_005131, ksc496i0h, 2,5-dibromo-4-methoxyanisole, 1,4-dibromo-3,6-dimethoxybenzene | |
| COC1=CC(=C(C=C1Br)OC)Br | |
| 295.958 | |
| 295.96 | |
| Crystalline Powder |
Chemical Identifiers
| 2674-34-2 | |
| 295.958 | |
| CHCLRVOURKGRSW-UHFFFAOYSA-N | |
| 231240 | |
| COC1=CC(=C(C=C1Br)OC)Br |
| C8H8Br2O2 | |
| MFCD00025694 | |
| 2,5-dibromo-1,4-dimethoxybenzene, 1,4-dibromo-2,5-dimethoxy-benzene, 2,5-dibromohydroquinone dimethyl ether, benzene, 1,4-dibromo-2,5-dimethoxy, pubchem16840, acmc-209gsv, maybridge1_005131, ksc496i0h, 2,5-dibromo-4-methoxyanisole, 1,4-dibromo-3,6-dimethoxybenzene | |
| 1,4-dibromo-2,5-dimethoxybenzene |
Safety and Handling
TSCA : No