missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2,5-dimethoxybenzene 98.0+%, TCI America™

Catalog No. p-7128023
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39665G 5 g
D396625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39665G Supplier TCI America Supplier No. D39665G

Chemical Identifiers

CAS 2674-34-2
Molecular Formula C8H8Br2O2
Molecular Weight (g/mol) 295.958
MDL Number MFCD00025694
InChI Key CHCLRVOURKGRSW-UHFFFAOYSA-N
Synonym 2,5-dibromo-1,4-dimethoxybenzene, 1,4-dibromo-2,5-dimethoxy-benzene, 2,5-dibromohydroquinone dimethyl ether, benzene, 1,4-dibromo-2,5-dimethoxy, pubchem16840, acmc-209gsv, maybridge1_005131, ksc496i0h, 2,5-dibromo-4-methoxyanisole, 1,4-dibromo-3,6-dimethoxybenzene
PubChem CID 231240
IUPAC Name 1,4-dibromo-2,5-dimethoxybenzene
SMILES COC1=CC(=C(C=C1Br)OC)Br

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 5 g
Formula Weight 295.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromo-2,5-dimethoxybenzene