missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2,5-diiodobenzene 96.0+%, TCI America™

Catalog No. p-7127986
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39045G 5 g
D390425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39045G Supplier TCI America Supplier No. D39045G

Chemical Identifiers

CAS 63262-06-6
Molecular Formula C6H2Br2I2
Molecular Weight (g/mol) 487.699
MDL Number MFCD00082898
InChI Key IVKPEQAIHJWGGT-UHFFFAOYSA-N
PubChem CID 11123771
IUPAC Name 1,4-dibromo-2,5-diiodobenzene
SMILES C1=C(C(=CC(=C1I)Br)I)Br

Specifications

Melting Point 167°C
Color White
Quantity 5 g
Formula Weight 487.70
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromo-2,5-diiodobenzene