accessibility menu, dialog, popup

UserName

IBrIBr

CAS RN 63262-06-6

IUPAC Name: 1,4-dibromo-2,5-diiodobenzene
Synonyms: 1,4-dibromo-2,5-diiodobenzene
Molecular Weight (g/mol): 487.7
Molecular Formula: C6H2Br2I2
InChi Key: IVKPEQAIHJWGGT-UHFFFAOYSA-N
SMILES: BrC1=CC(I)=C(Br)C=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results