missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2,5-difluorobenzene 98.0+%, TCI America™

Catalog No. D186410G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D186410G 10 g
D186425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D186410G Supplier TCI America Supplier No. D186410G

Chemical Identifiers

CAS 327-51-5
Molecular Formula C6H2Br2F2
Molecular Weight (g/mol) 271.887
MDL Number MFCD00000346
InChI Key GLVMLJCMUBZVTJ-UHFFFAOYSA-N
Synonym 1,4-dibromo-2,5-difluoro-benzene, benzene, 1,4-dibromo-2,5-difluoro, 2,5-difluoro-1,4-dibromo benzene, maybridge1_003946, pubchem3441, acmc-1cmi9, chemwish ic06783, ksc495q1l, emolecules 493137, 2,5-dibromo-1,4-difluorobenzene
PubChem CID 67596
IUPAC Name 1,4-dibromo-2,5-difluorobenzene
SMILES C1=C(C(=CC(=C1Br)F)Br)F

Specifications

Melting Point 65°C
Color White
Boiling Point 96°C
Quantity 10 g
Formula Weight 271.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromo-2,5-difluorobenzene