accessibility menu, dialog, popup

UserName

CAS RN 327-51-5

IUPAC Name: 1,4-dibromo-2,5-difluorobenzene
Synonyms: 1,4-dibromo-2,5-difluorobenzene
Molecular Weight (g/mol): 271.89
Molecular Formula: C6H2Br2F2
InChi Key: GLVMLJCMUBZVTJ-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(F)C=C1Br
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)