missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2,5-difluorobenzene, 98%

CAS: 327-51-5 | C6H2Br2F2 | 271.887 g/mol

Supplier:  Thermo Scientific Chemicals A1424814

 View more versions of this product

Catalog No. AAA1424814


Only null left
Explore available promotions

Description

Description

1,4-Dibromo-2,5-difluorobenzene has been used in the preparation of 1,2,4,5-tetrakis(phosphino)-3,6-difluorobenzenes and poly(2,5-difluoro-2,5-didodecyl-4,4-biphenylylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromo-2,5-difluorobenzene has been used in the preparation of 1,2,4,5-tetrakis(phosphino)-3,6-difluorobenzenes and poly(2,5-difluoro-2′,5′-didodecyl-4,4′-biphenylylene.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Stable under recommended storage conditions. Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
Specifications

Specifications

1,4-Dibromo-2,5-difluorobenzene
60°C to 64°C
C6H2Br2F2
25 g
1,4-dibromo-2,5-difluoro-benzene, benzene, 1,4-dibromo-2,5-difluoro, 2,5-difluoro-1,4-dibromo benzene, maybridge1_003946, pubchem3441, acmc-1cmi9, chemwish ic06783, ksc495q1l, emolecules 493137, 2,5-dibromo-1,4-difluorobenzene
GLVMLJCMUBZVTJ-UHFFFAOYSA-N
1,4-dibromo-2,5-difluorobenzene
67596
98%
327-51-5
96°C (20 mmHg)
MFCD00000346
2250095
Soluble in methanol. Insoluble in water.
C1=C(C(=CC(=C1Br)F)Br)F
271.887
271.89

Chemical Identifiers

327-51-5
271.887
GLVMLJCMUBZVTJ-UHFFFAOYSA-N
67596
C1=C(C(=CC(=C1Br)F)Br)F
C6H2Br2F2
MFCD00000346
1,4-dibromo-2,5-difluoro-benzene, benzene, 1,4-dibromo-2,5-difluoro, 2,5-difluoro-1,4-dibromo benzene, maybridge1_003946, pubchem3441, acmc-1cmi9, chemwish ic06783, ksc495q1l, emolecules 493137, 2,5-dibromo-1,4-difluorobenzene
1,4-dibromo-2,5-difluorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-317-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only