missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2,5-difluorobenzene, 98%

Catalog No. AAA1424822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1424822 100 g
AAA1424806 5 g
AAA1424814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1424822 Supplier Thermo Scientific Chemicals Supplier No. A1424822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 327-51-5 | C6H2Br2F2 | 271.887 g/mol

1,4-Dibromo-2,5-difluorobenzene has been used in the preparation of 1,2,4,5-tetrakis(phosphino)-3,6-difluorobenzenes and poly(2,5-difluoro-2,5-didodecyl-4,4-biphenylylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromo-2,5-difluorobenzene has been used in the preparation of 1,2,4,5-tetrakis(phosphino)-3,6-difluorobenzenes and poly(2,5-difluoro-2′,5′-didodecyl-4,4′-biphenylylene.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Stable under recommended storage conditions. Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 327-51-5
Molecular Formula C6H2Br2F2
Molecular Weight (g/mol) 271.887
MDL Number MFCD00000346
InChI Key GLVMLJCMUBZVTJ-UHFFFAOYSA-N
Synonym 1,4-dibromo-2,5-difluoro-benzene, benzene, 1,4-dibromo-2,5-difluoro, 2,5-difluoro-1,4-dibromo benzene, maybridge1_003946, pubchem3441, acmc-1cmi9, chemwish ic06783, ksc495q1l, emolecules 493137, 2,5-dibromo-1,4-difluorobenzene
PubChem CID 67596
IUPAC Name 1,4-dibromo-2,5-difluorobenzene
SMILES C1=C(C(=CC(=C1Br)F)Br)F

Specifications

Melting Point 60°C to 64°C
Boiling Point 96°C (20 mmHg)
Quantity 100 g
Beilstein 2250095
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 271.89
Percent Purity 98%
Chemical Name or Material 1,4-Dibromo-2,5-difluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-317-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only