missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromo-2,3-difluorobenzene 98.0+%, TCI America™
$82.02 - $279.60
Chemical Identifiers
| CAS | 156682-52-9 |
|---|---|
| Molecular Formula | C6H2Br2F2 |
| Molecular Weight (g/mol) | 271.887 |
| MDL Number | MFCD09835192 |
| InChI Key | RGXGEFSBDPGCEU-UHFFFAOYSA-N |
| PubChem CID | 18462393 |
| IUPAC Name | 1,4-dibromo-2,3-difluorobenzene |
| SMILES | C1=CC(=C(C(=C1Br)F)F)Br |
Chemical Identifiers
| 156682-52-9 | |
| 271.887 | |
| RGXGEFSBDPGCEU-UHFFFAOYSA-N | |
| 1,4-dibromo-2,3-difluorobenzene |
| C6H2Br2F2 | |
| MFCD09835192 | |
| 18462393 | |
| C1=CC(=C(C(=C1Br)F)F)Br |
Specifications
| 156682-52-9 | |
| MFCD09835192 | |
| RGXGEFSBDPGCEU-UHFFFAOYSA-N | |
| 1,4-dibromo-2,3-difluorobenzene | |
| 18462393 | |
| ≥98.0% (GC) | |
| 1,4-Dibromo-2,3-difluorobenzene |
| C6H2Br2F2 | |
| 1 g | |
| C1=CC(=C(C(=C1Br)F)F)Br | |
| 271.887 | |
| 271.89 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator