missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2,3-difluorobenzene 98.0+%, TCI America™

Catalog No. D49831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49831G 1 g
D49835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49831G Supplier TCI America Supplier No. D49831G
Only null left

Chemical Identifiers

CAS 156682-52-9
Molecular Formula C6H2Br2F2
Molecular Weight (g/mol) 271.887
MDL Number MFCD09835192
InChI Key RGXGEFSBDPGCEU-UHFFFAOYSA-N
PubChem CID 18462393
IUPAC Name 1,4-dibromo-2,3-difluorobenzene
SMILES C1=CC(=C(C(=C1Br)F)F)Br

Specifications

Quantity 1 g
Formula Weight 271.89
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dibromo-2,3-difluorobenzene
TSCA No
Recommended Storage Refrigerator