missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Di(1-pyrenyl)benzene 98.0+%, TCI America™
$266.56 - $799.68
Chemical Identifiers
| CAS | 475460-77-6 |
|---|---|
| Molecular Formula | C38H22 |
| Molecular Weight (g/mol) | 478.594 |
| MDL Number | MFCD12022457 |
| InChI Key | IJJYNFWMKNYNEW-UHFFFAOYSA-N |
| Synonym | 1,1′C-(1,4-Phenylene)bispyrene |
| PubChem CID | 23530003 |
| IUPAC Name | 1-(4-pyren-1-ylphenyl)pyrene |
| SMILES | C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C5=CC=C(C=C5)C6=C7C=CC8=CC=CC9=C8C7=C(C=C9)C=C6 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4922200MG
|
TCI America
D4922200MG |
200 mg |
Each for $266.56
|
|
|||||
|
D49221G
|
TCI America
D49221G |
1 g |
Each for $799.68
|
|
|||||
Chemical Identifiers
| 475460-77-6 | |
| 478.594 | |
| IJJYNFWMKNYNEW-UHFFFAOYSA-N | |
| 23530003 | |
| C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C5=CC=C(C=C5)C6=C7C=CC8=CC=CC9=C8C7=C(C=C9)C=C6 |
| C38H22 | |
| MFCD12022457 | |
| 1,1′C-(1,4-Phenylene)bispyrene | |
| 1-(4-pyren-1-ylphenyl)pyrene |
Specifications
| 475460-77-6 | |
| MFCD12022457 | |
| 1,1′C-(1,4-Phenylene)bispyrene | |
| C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C5=CC=C(C=C5)C6=C7C=CC8=CC=CC9=C8C7=C(C=C9)C=C6 | |
| 478.594 | |
| 478.59 | |
| Crystalline Powder |
| C38H22 | |
| 200 mg | |
| IJJYNFWMKNYNEW-UHFFFAOYSA-N | |
| 1-(4-pyren-1-ylphenyl)pyrene | |
| 23530003 | |
| ≥98.0% (HPLC) | |
| 1,4-Di(1-pyrenyl)benzene |
Safety and Handling
TSCA : No