CAS RN 475460-77-6
IUPAC Name:
1-[4-(pyren-1-yl)phenyl]pyrene
Synonyms:
1-[4-(pyren-1-yl)phenyl]pyrene
Molecular Weight (g/mol):
478.59
Molecular Formula:
C38H22
InChi Key:
IJJYNFWMKNYNEW-UHFFFAOYSA-N
SMILES:
C1=CC2=CC=C3C(=CC=C4C=CC(=C1)C2=C34)C1=CC=C(C=C1)C1=C2C=CC3=CC=CC4=CC=C(C=C1)C2=C34
1
–
1
of
1
results
1
1,4-Di(1-pyrenyl)benzene 98.0+%, TCI America™
CAS: 475460-77-6 Molecular Formula: C38H22 Molecular Weight (g/mol): 478.594 MDL Number: MFCD12022457 InChI Key: IJJYNFWMKNYNEW-UHFFFAOYSA-N Synonym: 1,1′C-(1,4-Phenylene)bispyrene PubChem CID: 23530003 IUPAC Name: 1-(4-pyren-1-ylphenyl)pyrene SMILES: C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C5=CC=C(C=C5)C6=C7C=CC8=CC=CC9=C8C7=C(C=C9)C=C6