missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Chrysenequinone 93.0+%, TCI America™
Specifications
Specifications
| Melting Point | 214°C |
| Quantity | 200 mg |
| Formula Weight | 258.28 |
| Percent Purity | ≥93.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,4-Chrysenequinone |
Safety and Handling
| TSCA | No |