accessibility menu, dialog, popup

UserName

OO

CAS RN 100900-16-1

IUPAC Name: 1,4-dihydrochrysene-1,4-dione
Synonyms: chrysene-1,4-dione
Molecular Weight (g/mol): 258.28
Molecular Formula: C18H10O2
InChi Key: UORKIKBNUWJNJF-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)C2=C3C=CC4=CC=CC=C4C3=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results