missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Thermo Scientific Chemicals

Catalog No. p-7056468
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH65397MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH65397MD Supplier Thermo Scientific Chemicals Supplier No. H65397MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 369-77-7 | Molecular Formula: C14H9Cl2F3N2O | Molecular Weight: 349.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 369-77-7
Molecular Formula C14H9Cl2F3N2O
Molecular Weight (g/mol) 349.13
MDL Number MFCD00867294
InChI Key ZFSXZJXLKAJIGS-UHFFFAOYSA-N
Synonym cloflucarban, halocarban, irgasan cf3, cloflucarbon, trifluoromethyldichlorocarbanilide, unii-i5zzy3dc5g, cloflucarban usan, 4,4'-dichloro-3-trifluoromethyl carbanilide, i5zzy3dc5g, halocarban inn
PubChem CID 9719
IUPAC Name 1-(4-chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
SMILES FC(F)(F)C1=CC(NC(=O)NC2=CC=C(Cl)C=C2)=CC=C1Cl

Specifications

Quantity 250 mg
Formula Weight 349.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-724-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only