CAS RN 369-77-7
IUPAC Name:
3-[4-chloro-3-(trifluoromethyl)phenyl]-1-(4-chlorophenyl)urea
Synonyms:
cloflucarban 卤卡班
Molecular Weight (g/mol):
349.13
Molecular Formula:
C14H9Cl2F3N2O
InChi Key:
ZFSXZJXLKAJIGS-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(NC(=O)NC2=CC=C(Cl)C=C2)=CC=C1Cl
1
–
1
of
1
results
1
Promotions Available
1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Thermo Scientific Chemicals
1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Thermo Scientific Chemicals
CAS: 369-77-7 Molecular Formula: C14H9Cl2F3N2O Molecular Weight (g/mol): 349.13 MDL Number: MFCD00867294 InChI Key: ZFSXZJXLKAJIGS-UHFFFAOYSA-N Synonym: cloflucarban,halocarban,irgasan cf3,cloflucarbon,trifluoromethyldichlorocarbanilide,unii-i5zzy3dc5g,cloflucarban usan,4,4'-dichloro-3-trifluoromethyl carbanilide,i5zzy3dc5g,halocarban inn PubChem CID: 9719 IUPAC Name: 1-(4-chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea SMILES: FC(F)(F)C1=CC(NC(=O)NC2=CC=C(Cl)C=C2)=CC=C1Cl